WHA230010060
Whatman® Benchkote® surface protector for ÄKTA® Pure
pack of 10 ea
Select a Size
$31.60
Select a Size
About This Item
$31.60
Recommended Products
material
(Absorbent paper and laminated polyethylene )
packaging
pack of 10 ea
manufacturer/tradename
Whatman 2300-10060
General description
- Cover the top buffer tray with Benchkote sheet and protect from buffer spillages and salt deposits.
- Simplify daily and weekly preventive maintenance of the top buffer tray of your ÄKTA systems Benchkote sheet
- Smooth, absorbent Whatman paper quickly absorbs spills.
- Laminated polyethylene layer prevents flow-through to the working surface.
- Fits the top buffer tray on ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems.
- Offered in three convenient pack sizes: 10/pk, 25/pk and 50/pk sheets.
- Convenient and easy to use.
Benchkote sheets are manufactured of absorbent,
impermeable material designed to protect lab surfaces against hazardous spills. The high-quality, smooth, and absorbent Whatman® paper, quickly absorbs liquid spills, and laminated polyethylene layer prevents flow-through to the working surface.
Features and Benefits
- Cover the top buffer tray with Benchkote sheet and protect from buffer spillages and salt deposits.
- Simplify daily and weekly preventive maintenance of the top buffer tray of your ÄKTA systems
- Smooth, absorbent Whatman paper quickly absorbs spills.
- Laminated polyethylene layer prevents flow-through to the working surface.
- Fits the top buffer tray on ÄKTA go, ÄKTA avant, ÄKTA pure and ÄKTA start protein purification systems.
- Offered in three convenient pack sizes: 10/pk, 25/pk and 50/pk sheets.
- Convenient and easy to use.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)