Documents
WHA10461000
Whatman® FP 025/1 filter holder
Synonym(s):
Z698504
About This Item
UNSPSC Code:
41104922
NACRES:
NB.24
sterility
non-sterile
packaging
pkg of 10 ea
manufacturer/tradename
Whatman 10461000
Whatman Article No. 28415718 (US reference)
General description
The syringe filter holder has a female Luer lock inlet and male Luer outlet compatible with typical syringes. Polysulfone housing can be autoclaved with membrane in place for applications requiring a sterile filter assembly.
Features and Benefits
  • Durable polysulfone plastic construction provides moderate chemical compatibility with lighter weight than stainless steel filter holders.
  • Polysulfone filter holders are often used for clarification, venting applications, particulate removal or collection from aqueous, air or gas streams.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva