Skip to Content
MilliporeSigma
All Photos(1)

Documents

W6918

Potassium palmitate-1-13C

API for Clinical Studies, ≥99 atom % 13C, ≥99% (CP)

Synonym(s):

Hexadecanoic acid-1-13C potassium salt, Palmitic acid-1-13C potassium salt, Potassium hexadecanoate-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)1413CO2K
CAS Number:
Molecular Weight:
295.51
MDL number:
UNSPSC Code:
12352116

grade

cGMP

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

mp

285 °C (dec.) (lit.)

mass shift

M+1

storage temp.

15-25°C

SMILES string

[K+].CCCCCCCCCCCCCCC[13C]([O-])=O

InChI

1S/C16H32O2.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18;/h2-15H2,1H3,(H,17,18);/q;+1/p-1/i16+1;

InChI key

MQOCIYICOGDBSG-OMVBPHMTSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service