![Potassium palmitate-1-13C API for Clinical Studies, ≥99 atom % 13C, ≥99% (CP)](/deepweb/assets/sigmaaldrich/product/structures/338/901/1bcfa039-602a-4b8b-9d50-252976d5252a/640/1bcfa039-602a-4b8b-9d50-252976d5252a.png)
Potassium palmitate-1-13C
![Potassium palmitate-1-13C API for Clinical Studies, ≥99 atom % 13C, ≥99% (CP)](/deepweb/assets/sigmaaldrich/product/structures/338/901/1bcfa039-602a-4b8b-9d50-252976d5252a/640/1bcfa039-602a-4b8b-9d50-252976d5252a.png)
Synonym(s):
Hexadecanoic acid-1-13C potassium salt, Palmitic acid-1-13C potassium salt, Potassium hexadecanoate-1-13C
Linear Formula:
CH3(CH2)1413CO2K
CAS Number:
Molecular Weight:
295.51
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3