Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W6667

D-Glucose-2-13C

API for Clinical Studies, ≥99 atom % 13C, ≥98% (CP)

Synonym(s):

Dextrose-2-13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC5H12O6
CAS Number:
Molecular Weight:
181.15
MDL number:
UNSPSC Code:
12352116

grade

cGMP

isotopic purity

≥99 atom % 13C

Assay

≥98% (CP)

optical activity

[α]25/D +52.0, c = 2 in H2O (trace NH4OH)

mp

150-152 °C (lit.)

mass shift

M+1

storage temp.

15-25°C

SMILES string

OC[C@H]1O[C@H](O)[13C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i5+1

InChI key

WQZGKKKJIJFFOK-UKLRSMCWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service