![D-Glucose-2-13C API for Clinical Studies, ≥99 atom % 13C, ≥98% (CP)](/deepweb/assets/sigmaaldrich/product/structures/264/570/811bc732-1789-4628-aeea-a4652d4f65a8/640/811bc732-1789-4628-aeea-a4652d4f65a8.png)
D-Glucose-2-13C
![D-Glucose-2-13C API for Clinical Studies, ≥99 atom % 13C, ≥98% (CP)](/deepweb/assets/sigmaaldrich/product/structures/264/570/811bc732-1789-4628-aeea-a4652d4f65a8/640/811bc732-1789-4628-aeea-a4652d4f65a8.png)
Synonym(s):
Dextrose-2-13C
Empirical Formula (Hill Notation):
13CC5H12O6
CAS Number:
Molecular Weight:
181.15
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Special Grade
Showing 1-3 of 3