Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W505102

Sigma-Aldrich

2,6-Diisopropylphenol

≥97%

Synonym(s):

2,6-Bis(1-methylethyl)phenol, 2,6-Bis(isopropyl)phenol, Propofol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2C6H3OH
CAS Number:
Molecular Weight:
178.27
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

vapor pressure

5.6 mmHg ( 100 °C)

Assay

≥97%

refractive index

n20/D 1.514 (lit.)

bp

256 °C/764 mmHg (lit.)

mp

18 °C (lit.)

density

0.962 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

SMILES string

CC(C)c1cccc(C(C)C)c1O

InChI

1S/C12H18O/c1-8(2)10-6-5-7-11(9(3)4)12(10)13/h5-9,13H,1-4H3

InChI key

OLBCVFGFOZPWHH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annette P N Kutter et al.
Anesthesia and analgesia, 121(1), 99-107 (2015-03-06)
In critically ill patients with significant pulmonary hypertension (PH), close perioperative cardiovascular monitoring is mandatory, considering the increased morbidity and mortality in this patient group. Although the pulmonary artery catheter is still the standard for the diagnosis of PH, its
Michael Schwarzl et al.
American journal of physiology. Heart and circulatory physiology, 309(9), H1407-H1418 (2015-09-06)
Heart failure with preserved ejection fraction (HFPEF) evolves with the accumulation of risk factors. Relevant animal models to identify potential therapeutic targets and to test novel therapies for HFPEF are missing. We induced hypertension and hyperlipidemia in landrace pigs (n
Philippe E Dubois et al.
European journal of anaesthesiology, 31(8), 404-410 (2013-07-26)
Disturbances in the thumb's movement interfere with the functioning of acceleromyography in many clinical settings. The short and light (SL) train-of-four (TOF)-Tube is a new version of a rigid tubular device that was designed to protect the thumb from external
Kim van Loon et al.
Anesthesia and analgesia, 119(1), 49-55 (2014-05-20)
Propofol, a short-acting hypnotic drug, is increasingly administered by a diverse group of specialists (e.g., cardiologists, gastroenterologists) during diagnostic and therapeutic procedures. Standard monitoring during sedation comprises continuous pulse oximetry with visual assessment of the patient's breathing pattern. Because undetected
Felix Schlegel et al.
NeuroImage, 116, 40-49 (2015-05-17)
In recent years, the number of functional MRI (fMRI) studies in mice has been rapidly increasing. Technological improvements provide the sensitivity required to match the high demands on spatial and temporal resolution and to analyze fast and small signal components

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service