Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W403326

Sigma-Aldrich

1,3-Dihydroxyacetone dimer

97%

Synonym(s):

2,5-Dihydroxydioxane-2,5-dimethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
112910
MDL number:
UNSPSC Code:
12164502
Flavis number:
13.11

biological source

synthetic

Assay

97%

form

powder

color

white to off-white

mp

75-80 °C (lit.)

SMILES string

OCC1(O)COC(O)(CO)CO1

InChI

1S/C6H12O6/c7-1-5(9)3-12-6(10,2-8)4-11-5/h7-10H,1-4H2

InChI key

KEQUNHIAUQQPAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Component of tanning products

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service