Skip to Content
MilliporeSigma
1,3-Dihydroxyacetone dimer 97%

1,3-Dihydroxyacetone dimer

1,3-Dihydroxyacetone dimer 97%
Synonym(s):
2,5-Dihydroxydioxane-2,5-dimethanol
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing