Skip to Content
MilliporeSigma
All Photos(1)

Documents

W393411

Sigma-Aldrich

cis-3-Hexenyl pyruvate

≥93%

Synonym(s):

3-Hexenyl 2-oxopropanoate, (Z)-3-Hexen-1-yl pyruvate, (Z)-3-Hexenyl 2-oxopropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O3
CAS Number:
Molecular Weight:
170.21
FEMA Number:
3934
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.565
NACRES:
NA.21

reg. compliance

FDA 21 CFR 117

Quality Level

Assay

≥93%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

banana; cucumber; green; melon; oily

SMILES string

CC\C=C/CCOC(=O)C(C)=O

InChI

1S/C9H14O3/c1-3-4-5-6-7-12-9(11)8(2)10/h4-5H,3,6-7H2,1-2H3/b5-4-

InChI key

LKNXTZXOBHAYSR-PLNGDYQASA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service