Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W404411

Sigma-Aldrich

Geranyl tiglate

≥95%, FG

Synonym(s):

Geranyl 2-methylcrotonate

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H24O2
CAS Number:
Molecular Weight:
236.35
FEMA Number:
4044
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.383
NACRES:
NA.21
grade:
FG
Halal
Kosher
food allergen:
no known allergens

grade

FG
Halal
Kosher

Quality Level

reg. compliance

EU 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

refractive index

n20/D 1.480-1.484 (lit.)

bp

149-151 °C/7 mmHg (lit.)

density

0.923 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

geranium; fruity; herbaceous; sweet

SMILES string

[H]\C(C)=C(\C)C(=O)OC\C=C(/C)CC\C=C(\C)C

InChI

1S/C15H24O2/c1-6-14(5)15(16)17-11-10-13(4)9-7-8-12(2)3/h6,8,10H,7,9,11H2,1-5H3/b13-10+,14-6+

InChI key

OGHBUHJLMHQMHS-KRDNBFTESA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Navigate halal certification, food & beverage compliance, and global standards (BPJPH, SMIIC, JAKIM) with our halal products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service