Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W374008

Sigma-Aldrich

Anisyl phenylacetate

≥97%

Synonym(s):

4-Methoxybenzyl phenylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CO2CH2C6H4OCH3
CAS Number:
Molecular Weight:
256.30
FEMA Number:
3740
EC Number:
Council of Europe no.:
233
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.706
NACRES:
NA.21
Organoleptic:
anise
grade:
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

Halal

Assay

≥97%

refractive index

n20/D 1.559 (lit.)

bp

370 °C (lit.)

density

1.128 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

anise

SMILES string

COc1ccc(COC(=O)Cc2ccccc2)cc1

InChI

1S/C16H16O3/c1-18-15-9-7-14(8-10-15)12-19-16(17)11-13-5-3-2-4-6-13/h2-10H,11-12H2,1H3

InChI key

VCYWCSZLXMMLLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service