Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W299618

Sigma-Aldrich

Rum ether

Synonym(s):

Ethyl oxyhydrate

Sign Into View Organizational & Contract Pricing

Select a Size

W299618-SAMPLE-K
$74.30
1 KG
$102.00
8 KG
$318.00
4 KG
$338.00

$74.30


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W299618-SAMPLE-K
$74.30
1 KG
$102.00
8 KG
$318.00
4 KG
$338.00

About This Item

CAS Number:
FEMA Number:
2996
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
Kosher
biological source:
synthetic
food allergen:
no known allergens

$74.30


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

Kosher

reg. compliance

FDA 21 CFR 172.515

refractive index

n20/D 1.366 (lit.)

bp

76 °C (lit.)

density

0.825 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

whiskey; wine-like

Looking for similar products? Visit Product Comparison Guide

General description

Rum ether is a complex mixture of volatile substances that can be used as a flavoring agent for its rum-like odor/flavor.[1]

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scientific Opinion of Flavouring Group Evaluation 500 (FGE. 500): rum ether.
Silano V, et al.
EFSA Journal, 15(8), e04897-e04897 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service