Skip to Content
MilliporeSigma
All Photos(1)

Documents

W367000

Sigma-Aldrich

2,4-Dodecadienal

natural (US), 10 wt. % in medium chain triglycerides (MCTG)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20O
CAS Number:
Molecular Weight:
180.29
FEMA Number:
3670
Beilstein:
1776172
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

botanical

grade

Halal
Kosher
natural (US)

concentration

10 wt. % in medium chain triglycerides (MCTG)

density

0.865 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

vegetable

SMILES string

CCCCCCC\C=C\C=C\C=O

InChI

1S/C12H20O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h8-12H,2-7H2,1H3/b9-8+,11-10+

InChI key

QKTZBZWNADPFOL-BNFZFUHLSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.0 °F - closed cup

Flash Point(C)

93.89 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service