![2,4-Dodecadienal natural (US), 10 wt. % in medium chain triglycerides (MCTG)](/deepweb/assets/sigmaaldrich/product/structures/720/516/78057c42-1b86-493b-9629-29c857b0f74a/640/78057c42-1b86-493b-9629-29c857b0f74a.png)
2,4-Dodecadienal
![2,4-Dodecadienal natural (US), 10 wt. % in medium chain triglycerides (MCTG)](/deepweb/assets/sigmaaldrich/product/structures/720/516/78057c42-1b86-493b-9629-29c857b0f74a/640/78057c42-1b86-493b-9629-29c857b0f74a.png)
Empirical Formula (Hill Notation):
C12H20O
CAS Number:
Molecular Weight:
180.29
Products
Product Category
Brand
Biological Source
Formula Weight
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing