Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W361801

Sigma-Aldrich

5-Phenyl-1-pentanol

≥98%

Synonym(s):

Benzenepentanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)5OH
CAS Number:
Molecular Weight:
164.24
FEMA Number:
3618
Beilstein:
1935233
EC Number:
Council of Europe no.:
674
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.051

biological source

synthetic

grade

Kosher

Assay

≥98%

refractive index

n20/D 1.516 (lit.)

bp

155 °C/20 mmHg (lit.)

density

0.975 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

SMILES string

OCCCCCc1ccccc1

InChI

1S/C11H16O/c12-10-6-2-5-9-11-7-3-1-4-8-11/h1,3-4,7-8,12H,2,5-6,9-10H2

InChI key

DPZMVZIQRMVBBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hidetaka Nagatomo et al.
Bioscience, biotechnology, and biochemistry, 69(1), 128-136 (2005-01-25)
Hyperthermostable beta-glucosidase from Pyrococcus furiosus was enclosed in gelatin gel by cross-linking with transglutaminase. Gelatin-immobilized beta-glucosidase was considerably more thermostable than the native enzyme. Lyophilized immobilisate was stored at 90 degrees C for 1 month without loss of activity. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service