Skip to Content
MilliporeSigma
5-Phenyl-1-pentanol 98%

5-Phenyl-1-pentanol

5-Phenyl-1-pentanol 98%
Synonym(s):
Benzenepentanol
Linear Formula:
C6H5(CH2)5OH
CAS Number:
Molecular Weight:
164.24
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing