Skip to Content
MilliporeSigma
All Photos(1)

Documents

W323802

Sigma-Aldrich

2,2′-(Thiodimethylene)difuran

≥98%

Synonym(s):

Furfuryl sulfide, Difurfuryl monosulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2S
CAS Number:
Molecular Weight:
194.25
FEMA Number:
3238
EC Number:
Council of Europe no.:
11438
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.056
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Halal

Agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 117

Assay

≥98%

refractive index

n20/D 1.556 (lit.)

bp

135-143 °C/14 mmHg (lit.)

mp

29-34 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

coffee; meaty; earthy; roasted

SMILES string

C(SCc1ccco1)c2ccco2

InChI

1S/C10H10O2S/c1-3-9(11-5-1)7-13-8-10-4-2-6-12-10/h1-6H,7-8H2

InChI key

UYLKDZXJEKFFHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 2.5 ppm

Other Notes

Natural occurrence: Coffee.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 26, 71-71 (2001)
Mosciano, G.
Perfumer & Flavorist, 27(4), 68-68 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service