Skip to Content
MilliporeSigma
2,2′-(Thiodimethylene)difuran ≥98%

2,2′-(Thiodimethylene)difuran

2,2′-(Thiodimethylene)difuran ≥98%
Synonym(s):
Furfuryl sulfide, Difurfuryl monosulfide
Empirical Formula (Hill Notation):
C10H10O2S
CAS Number:
Molecular Weight:
194.25
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing