Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W304905

Sigma-Aldrich

Terpinyl butyrate, mixture of isomers

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24O2
CAS Number:
Molecular Weight:
224.34
FEMA Number:
3049
EC Number:
Council of Europe no.:
278
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.052
NACRES:
NA.21

biological source

synthetic

Assay

≥95%

optical activity

[α]20/D −9.0°, neat

refractive index

n20/D 1.4650 (lit.)

bp

244 °C (lit.)

density

0.938 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; balsamic

SMILES string

CCCC(=O)OC(C)(C)C1CCC(C)=CC1

InChI

1S/C14H24O2/c1-5-6-13(15)16-14(3,4)12-9-7-11(2)8-10-12/h7,12H,5-6,8-10H2,1-4H3

InChI key

LWKWNIYBQLKBMQ-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service