Skip to Content
MilliporeSigma
All Photos(1)

Documents

W251208

Sigma-Aldrich

Geranyl butyrate

≥95%, FCC

Synonym(s):

Butyric acid geranyl ester, Trans-3,7-dimethyl-2,6-octadien-1-yl butyrate, [(2E)-3,7-dimethylocta-2,6-dienyl] butanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
224.34
FEMA Number:
2512
EC Number:
Council of Europe no.:
274
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.048
NACRES:
NA.21

grade

Kosher

Quality Level

reg. compliance

FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

impurities

<35% citronellyl and neryl butyrates

refractive index

n20/D 1.461 (lit.)

bp

151-153 °C/18 mmHg (lit.)

density

0.896 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; fruity; waxy; rose; sweet

SMILES string

[H]\C(CC\C(C)=C(/[H])COC(=O)CCC)=C(/C)C

InChI

1S/C14H24O2/c1-5-7-14(15)16-11-10-13(4)9-6-8-12(2)3/h8,10H,5-7,9,11H2,1-4H3/b13-10+

InChI key

ZSBOMYJPSRFZAL-JLHYYAGUSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service