Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W295817

Sigma-Aldrich

Propyl propionate

natural (US), ≥98%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COOCH2CH2CH3
CAS Number:
Molecular Weight:
116.16
FEMA Number:
2958
Beilstein:
1699993
EC Number:
Council of Europe no.:
403c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.122

grade

FG
Halal
Kosher
natural (US)

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor density

4 (vs air)

Assay

≥98%

refractive index

n20/D 1.393 (lit.)

bp

122-124 °C (lit.)

mp

−76 °C (lit.)

density

0.881 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

wine-like

SMILES string

CCCOC(=O)CC

InChI

1S/C6H12O2/c1-3-5-8-6(7)4-2/h3-5H2,1-2H3

InChI key

MCSINKKTEDDPNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F - closed cup - Information taken from reference works and the literature.

Flash Point(C)

24 °C - closed cup - Information taken from reference works and the literature.

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service