Skip to Content
MilliporeSigma
Propyl propionate ≥98%, FCC, FG

Propyl propionate

Propyl propionate ≥98%, FCC, FG
Linear Formula:
CH3CH2COOCH2CH2CH3
CAS Number:
Molecular Weight:
116.16
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing