Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W290807

Sigma-Aldrich

Piperidine

≥99%, FCC

Synonym(s):

Hexahydropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11N
CAS Number:
Molecular Weight:
85.15
FEMA Number:
2908
Beilstein:
102438
EC Number:
Council of Europe no.:
675
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.010

biological source

synthetic

grade

Halal
Kosher

reg. compliance

FCC
FDA 21 CFR 172.515

vapor density

3 (vs air)

vapor pressure

23 mmHg ( 20 °C)
63.6 mmHg ( 37.8 °C)

Assay

≥99%

form

liquid

refractive index

n20/D 1.452 (lit.)

bp

106 °C (lit.)

mp

−13 °C (lit.)

density

0.862 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; floral; sweet

SMILES string

C1CCNCC1

InChI

1S/C5H11N/c1-2-4-6-5-3-1/h6H,1-5H2

InChI key

NQRYJNQNLNOLGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

GMO free

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of (-)-kopsinine by an asymmetric one-pot [n+2+3] cyclization.
Shingo Harada et al.
Chemistry, an Asian journal, 7(10), 2196-2198 (2012-08-22)
J Phillip Turner et al.
ACS chemical neuroscience, 5(7), 552-558 (2014-04-03)
Alzheimer's disease (AD) is the most common form of dementia and the sixth leading cause of death in the United States. Plaques composed of aggregated amyloid-beta protein (Aβ) accumulate between the neural cells in the brain and are associated with
Marek Łuczkowski et al.
Inorganic chemistry, 47(23), 10875-10888 (2008-10-31)
A de novo protein design strategy provides a powerful tool to elucidate how heavy metals interact with proteins.Cysteine derivatives of the TRI peptide family (Ac-G(LKALEEK)4G-NH2) have been shown to bind heavy metals in an unusual trigonal geometry. Our present objective
Xuwang Chen et al.
Bioorganic & medicinal chemistry, 20(12), 3856-3864 (2012-05-18)
A novel series of piperidine-linked amino-triazine derivatives were designed, synthesized and evaluated for in vitro anti-HIV activity as non-nucleoside reverse transcriptase inhibitors on the basis of our previous work. Screening results indicated that most compounds showed excellent activity against wild-type
Elisabeth T Hennessy et al.
Science (New York, N.Y.), 340(6132), 591-595 (2013-05-04)
The manipulation of traditionally unreactive functional groups is of paramount importance in modern chemical synthesis. We have developed an iron-dipyrrinato catalyst that leverages the reactivity of iron-borne metal-ligand multiple bonds to promote the direct amination of aliphatic C-H bonds. Exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service