MilliporeSigma
  • Complex N-heterocycle synthesis via iron-catalyzed, direct C-H bond amination.
Complex N-heterocycle synthesis via iron-catalyzed, direct C-H bond amination.
Science (New York, N.Y.) (2013-05-04)
Elisabeth T Hennessy, Theodore A Betley
ABSTRACT
The manipulation of traditionally unreactive functional groups is of paramount importance in modern chemical synthesis. We have developed an iron-dipyrrinato catalyst that leverages the reactivity of iron-borne metal-ligand multiple bonds to promote the direct amination of aliphatic C-H bonds. Exposure of organic azides to the iron dipyrrinato catalyst furnishes saturated, cyclic amine products (N-heterocycles) bearing complex core-substitution patterns. This study highlights the development of C-H bond functionalization chemistry for the formation of saturated, cyclic amine products and should find broad application in the context of both pharmaceuticals and natural product synthesis.
Sigma-Aldrich
Piperidine solution, suitable for peptide synthesis, 20% in DMF
Sigma-Aldrich
Azetidine, 98%
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
Piperidine, puriss. p.a., ≥99.0% (GC/T)
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Piperidine, biotech. grade, ≥99.5%