Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W289205

Sigma-Aldrich

2-Phenylpropyl isobutyrate

≥97%, FG

Synonym(s):

Hydratropyl isobutyrate

Sign Into View Organizational & Contract Pricing

Select a Size

W289205-SAMPLE-K
$112.00
100 G
$537.00

$112.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W289205-SAMPLE-K
$112.00
100 G
$537.00

About This Item

Linear Formula:
(CH3)2CHCO2CH2CH(C6H5)CH3
CAS Number:
Molecular Weight:
206.28
FEMA Number:
2892
EC Number:
Council of Europe no.:
2087
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.485
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

$112.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥97%

refractive index

n20/D 1.485 (lit.)

density

0.973 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; floral

SMILES string

CC(C)C(=O)OCC(C)c1ccccc1

InChI

1S/C13H18O2/c1-10(2)13(14)15-9-11(3)12-7-5-4-6-8-12/h4-8,10-11H,9H2,1-3H3

InChI key

LZTCJUAHYXNKRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service