Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W280110

Sigma-Aldrich

2-Octanol

natural, ≥97%, FG

Synonym(s):

sec-Caprylic alcohol, Capryl alcohol, Ethyl-N-amylcarbinol, Ethylpentylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(OH)CH3
CAS Number:
Molecular Weight:
130.23
FEMA Number:
2801
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.022
NACRES:
NA.21
Organoleptic:
green; spicy; earthy; woody
grade:
FG
Kosher
natural
food allergen:
no known allergens

grade

FG
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 110
FDA 21 CFR 172.515
FDA 21 CFR 176.180
FDA 21 CFR 176.210

Assay

≥97%

refractive index

n20/D 1.426 (lit.)
n20/D 1.426

bp

174-181 °C (lit.)

density

0.819 g/mL at 25 °C (lit.)
0.819 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; spicy; earthy; woody

SMILES string

CCCCCCC(C)O

InChI

1S/C8H18O/c1-3-4-5-6-7-8(2)9/h8-9H,3-7H2,1-2H3

InChI key

SJWFXCIHNDVPSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rong Zhang et al.
Journal of agricultural and food chemistry, 62(33), 8302-8307 (2014-07-30)
Nonvolatile compounds play important roles in the quality of alcoholic beverages. In our previous work, a type of cyclooctapeptide lichenysin was newly identified in Chinese strong-aroma type liquor. In this work, it was found that lichenysin could selectively affect aroma
Anzi Wang et al.
Journal of chromatography. A, 1374, 216-223 (2014-12-06)
Fabrication technologies for microelectromechanical systems (MEMS) allow miniaturization of conventional benchtop gas chromatography (GC) to portable, palm-sized microfabricated GC (μGC) devices, which are suitable for on-site chemical analysis and remote sensing. The separation performance of μGC systems, however, has not
Xu Wang et al.
Food chemistry, 182, 41-46 (2015-04-07)
The volatile compounds in gingko wine, a novel functional wine, were extracted by head-space solid phase micro-extraction (SPME) and analyzed by gas chromatography-mass spectrometry (GC-MS) coupled with odor activity value (OAV) and relative odor contribution (ROC) analyses. In addition, the
Pankaj K Sethi et al.
Journal of pharmacological and toxicological methods, 70(1), 106-111 (2014-06-15)
A simple, reliable procedure was developed to measure binding of pyrethroid insecticides to total proteins and lipoproteins of rat and human plasma. The extent of binding of (14)C-labeled deltamethrin (DLM), cis-permethrin (CIS) and trans-permethrin (TRANS) was quantified by a 3-step
Navideh Sadoughi et al.
Journal of agricultural and food chemistry, 63(11), 2877-2885 (2015-02-24)
An optimized method for the quantitation of volatile compounds responsible for off-aromas, such as earthy odors, found in wine and grapes was developed. The method involved a fast and simple headspace solid-phase microextraction-gas chromatography-mass spectrometry (HS-SPME-GC-MS) for simultaneous determination of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service