Skip to Content
MilliporeSigma
2-Octanol 97%

2-Octanol

2-Octanol 97%
Synonym(s):
sec-Caprylic alcohol, Capryl alcohol, Ethyl-N-amylcarbinol, Ethylpentylcarbinol, 2-Octanol
Linear Formula:
CH3(CH2)5CH(OH)CH3
CAS Number:
Molecular Weight:
130.23

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing