Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

STS0014

Sigma-Aldrich

TERGITOL 15-S-15

greener alternative

Non-ionic surfactant

Synonym(s):

Alcohols, C12-14-secondary, ethoxylated, Secondary alcohol ethoxylate 15 EO

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$69.80
500 G
$88.20

$69.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$69.80
500 G
$88.20

About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.28

$69.80


Please contact Customer Service for Availability

Request a Bulk Order

grade

laboratory grade

Quality Level

description

non-ionic

form

solid

mol wt

average mol wt 860 g/mol by calculation

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

hybridization: suitable

CMC

162 ppm (25°C)

viscosity

43 cP(25 °C)

transition temp

cloud point >100 °C (1 wt% actives aqueous solution)
pour point 29 °C

solubility

water: freely soluble (some compositions may form gels)

density

1.007 g/mL at 20 °C

HLB

15.4

greener alternative category

General description

Tergitol 15-S-15 is a secondary alcohol ethoxylate with 15 ethylene oxide units
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".

Features and Benefits

  • High HLB emulsifier & dispersant
  • Provides freeze/thaw & ionic stability
  • Electrolyte solubility
  • High temperature wetting
  • Readily biodegradable

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

449.6 °F - closed cup

Flash Point(C)

232 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service