Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W241806

Sigma-Aldrich

Ethyl acrylate

≥99.5%, stabilized, FG

Synonym(s):

Ethyl 2-propenoate, Acrylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCOOC2H5
CAS Number:
Molecular Weight:
100.12
FEMA Number:
2418
Beilstein:
773866
EC Number:
Council of Europe no.:
245
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.037
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

3.5 (vs air)

vapor pressure

31 mmHg ( 20 °C)

Assay

≥99.5%

autoignition temp.

721 °F

contains

10-30 ppm MEHQ as stabilizer (synthetic)

expl. lim.

12.1 %

refractive index

n20/D 1.406 (lit.)

bp

99 °C (lit.)

mp

−71 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

plastic

SMILES string

CCOC(=O)C=C

InChI

1S/C5H8O2/c1-3-5(6)7-4-2/h3H,1,4H2,2H3

InChI key

JIGUQPWFLRLWPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl acrylate can be used as a flavoring agent in the food industry. It is one of the main volaitle components identified in organic passion fruit pulp.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of the cultivation system in the aroma of the volatile compounds and total antioxidant activity of passion fruit
Janzantti NS, et al.
LWT--Food Science and Technology, 46(2), 511-518 (2012)
Burdock, GA
Encyclopedia of Food and Color Additives, 1, 945-946 (1997)
Julia Kredel et al.
Polymers, 11(12) (2020-01-01)
Fluoropolymers represent a unique class of functional polymers due to their various interesting and important properties such as thermal stability, resistance toward chemicals, repellent behaviors, and their low refractive indices in comparison to other polymeric materials. Based on the latter
Marilene S Oliveira et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 10(10), 1546-1555 (2011-07-08)
Singlet molecular oxygen O(2)((1)Δ(g)) is a potent oxidant that can react with different biomolecules, including DNA, lipids and proteins. Many polycyclic aromatic hydrocarbons have been studied as O(2)((1)Δ(g)) chemical traps. Nevertheless, a suitable modification in the polycyclic aromatic ring must
J C O Villanova et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 79(3), 664-673 (2011-09-29)
The design of new excipients that extend the release of drugs from tablets over prolonged periods is essential in reaching enhanced therapeutic performances. In this sense, the objective of this study was to develop new excipients, based on acrylic monomers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service