Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00578

Sigma-Aldrich

Methacrylic acid

(stabilised with hydroquinone monomethyl ether) for synthesis

Synonym(s):

Methacrylic acid, 2-Methylpropenoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$47.70
500 ML
$63.10
2.5 L
$140.00
25 KG
$824.00

$47.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$47.70
500 ML
$63.10
2.5 L
$140.00
25 KG
$824.00

About This Item

Linear Formula:
CH2C(CH3)COOH
CAS Number:
Molecular Weight:
86.09
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-204-4
NACRES:
NA.22

$47.70


Please contact Customer Service for Availability

Request a Bulk Order

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

<1 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

370 °C

potency

1320 mg/kg LD50, oral (Rat)

expl. lim.

1.6-8.7 % (v/v)

pH

2.0-2.2 (20 °C, 100 g/L in H2O)

bp

161 °C/1013 hPa

mp

15 °C

transition temp

flash point 65 °C

density

1.015 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

OC(=O)C(=C)C

InChI

1S/C4H6O2/c1-3(2)4(5)6/h1H2,2H3,(H,5,6)

InChI key

CERQOIWHTDAKMF-UHFFFAOYSA-N

Related Categories

General description

Methacrylicacid (MAA) is a liquid carboxylic acid that is colorless, viscous, and solublein most organic solvents. It is generated on a large scale in industrialapplications as a precursor to its esters, particularly methyl methacrylate(MMA).

Application

Methacrylic acid is used in the graft copolymerization with jute fiber.[1]

Analysis Note

Assay (GC, area%): ≥ 99 %
Density (d 20 °C/ 4 °C): 1.013 - 1.015
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service