Skip to Content
MilliporeSigma
All Photos(1)

Documents

W215902

Sigma-Aldrich

Bornyl acetate

≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29
FEMA Number:
2159
EC Number:
Council of Europe no.:
207
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.017

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

optical activity

[α]20/D −41°, neat

refractive index

n20/D 1.4635 (lit.)

bp

228-231 °C (lit.)

density

0.985 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody; sweet

SMILES string

CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C

InChI

1S/C12H20O2/c1-8(13)14-10-7-9-5-6-12(10,4)11(9,2)3/h9-10H,5-7H2,1-4H3/t9-,10+,12+/m0/s1

InChI key

KGEKLUUHTZCSIP-HOSYDEDBSA-N

Legal Information

For R&D Use and/or Food Manufacturing only

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

192.0 °F - closed cup

Flash Point(C)

88.89 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eri Matsubara et al.
Biomedical research (Tokyo, Japan), 32(2), 151-157 (2011-05-10)
(-)-Bornyl acetate is the main volatile constituent in numerous conifer oils and has a camphoraceous, pine-needle-like odor. It is frequently used as the conifer needle composition in soap, bath products, room sprays, and pharmaceutical products. However, the psychophysiological effects of
Leo A Paquette et al.
The Journal of organic chemistry, 68(18), 6905-6918 (2003-08-30)
An intramolecular anionic oxy-Cope rearrangement (44 --> 46) serves as the key step in a synthetic approach to the insect antifeedant dumsin. Initial investigations clarified the manner in which (--)-bornyl acetate may be transformed into the exo-norbornenol 44. Two routes
T Hirata et al.
Phytochemistry, 55(3), 197-202 (2001-01-06)
A peroxidase was purified from the culture medium of a suspension culture of Marchantia polymorpha (liverwort) after treatment with bornyl acetate, which acts as a chemical stress agent to the cells. The peroxidase was characterised as a glycoprotein of molecular
Xiaosong Wu et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 28(6), 505-507 (2005-10-08)
To study the analgesic effect and mechanism of bornyl acetate, the main ingredient of Amomum Villosum Volatile oil. The analgesic effects were tested by pressing tail method. The I, and II phase pain were observed with the pain model caused
E Tachikawa et al.
Biochemical pharmacology, 60(3), 433-440 (2000-06-17)
The crude extract of magnolia bark, an herbal drug, inhibited the secretion of catecholamines from bovine adrenal chromaffin cells stimulated by acetylcholine (ACh) in a concentration-dependent manner (200-900 microg/mL). The extract also diminished the secretion induced by high K(+), which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service