MilliporeSigma
  • (-)-Bornyl acetate induces autonomic relaxation and reduces arousal level after visual display terminal work without any influences of task performance in low-dose condition.
(-)-Bornyl acetate induces autonomic relaxation and reduces arousal level after visual display terminal work without any influences of task performance in low-dose condition.
Biomedical research (Tokyo, Japan) (2011-05-10)
Eri Matsubara, Mio Fukagawa, Tsuyoshi Okamoto, Koichiro Ohnuki, Kuniyoshi Shimizu, Ryuichiro Kondo
ABSTRACT
(-)-Bornyl acetate is the main volatile constituent in numerous conifer oils and has a camphoraceous, pine-needle-like odor. It is frequently used as the conifer needle composition in soap, bath products, room sprays, and pharmaceutical products. However, the psychophysiological effects of (-)-bornyl acetate remained unclear. We investigated the effects of breathing air mixed with (-)-bornyl acetate at different doses (low-dose and high-dose conditions) on the individuals during and after VDT (visual display terminal) work using a visual discrimination task. The amounts of (-)-bornyl acetate through our odorant delivery system for 40 min were 279.4 µg in the low-dose and 716.3 µg in the high-dose (-)-bornyl acetate condition. (-)-Bornyl acetate induced changes of autonomic nervous system for relaxation and reduced arousal level after VDT work without any influences of task performance in low-dose condition, but not in high-dose condition.