Documents
TEM-LC300AUCC
Sigma-Aldrich
Continuous Ultrathin Carbon Film Coated Lacey Carbon Supported Gold Grid
size 300 mesh, box of 5
Synonym(s):
LACEY CARBON FILM - AU 300 MESH 50 U 5/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 5
size
300 mesh
General description
Lacey Carbon Film Coated with a Continuous Layer of Ultrathin Carbon Film. Gold Grid, 300 mesh. The continuous ultrathin film on lacey film allows for the thinnest support film that still has adequate strength to withstand specimen preparation. The film (less than 3nm thick) lies across a carbon lacey film supported by a 300 mesh grid. The size of the holes in the lacey film differ widely from batch to batch but are generally in the range of ¼ μm to 5μm, which gives the equivalent support of at least 6000 mesh grid. Specimen material lying over the covered holes can be imaged in the TEM with practically no interference from the carbon film supporting it. This product is ideal for looking at nanotubes, virus particles and other small particulate material.