Key Documents
TEM-CF400HCU50
Sigma-Aldrich
Carbon Film Supported Copper Hexagonal Mesh Grid
size 400 mesh, box of 50
Synonym(s):
CARBON FILM ONLY ON 400 HEX COPPER GRIDS 50/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-CF400HCU50 is not currently sold in your country. Contact Technical Service
material
carbon film
copper grid
description
film layer thickness 5 - 6 nm carbon
packaging
box of 50
size
400 mesh
General description
A 5-6 nm thin film of pure carbon supported hexagonal mesh grids. Copper grid, 400 mesh. Carbon film show better electrical and heat conduction in the electron beam.
Choose from one of the most recent versions:
Certificates of Analysis (COA)