Documents
TEM-CF200AU
Sigma-Aldrich
Carbon Film Supported Gold Square Mesh
size 200 mesh, box of 25
Synonym(s):
CARBON FILM 200# GOLD 25/PK
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25
size
200 mesh
General description
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Gold grid, 200 mesh.