Key Documents
TEM-76042-41
Sigma-Aldrich
Micronporous Silicon Nitride TEM Window Grids for Cryo-EM Suspension
50nm Thick, pack of 10
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-76042-41 is not currently sold in your country. Contact Technical Service
packaging
pack of 10
General description
Silicon nitrode microporous TEM window with single square of 500x500 micron size, and 50 nm thick SiN. Plasma cleanable, less than 0.5nm variation in film thickness, tolerate temperatures above 1000 ˚C, provides flat, insulating and hydrophobic surfaces
Choose from one of the most recent versions:
Certificates of Analysis (COA)