Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T13404

Sigma-Aldrich

5,6,7,8-Tetrahydro-1-naphthol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O
CAS Number:
Molecular Weight:
148.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

264-265 °C/705 mmHg (lit.)

mp

69-71 °C (lit.)

SMILES string

Oc1cccc2CCCCc12

InChI

1S/C10H12O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h3,5,7,11H,1-2,4,6H2

InChI key

SCWNNOCLLOHZIG-UHFFFAOYSA-N

Gene Information

human ... BAD(572)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas B Senese et al.
Neuropharmacology, 141, 296-304 (2018-09-07)
A single base mutation in the Gαi2 protein (G184S) renders this Gα subunit insensitive to the negative modulatory effects of Regulator of G-protein Signaling (RGS) proteins. Mice expressing this RGS insensitive (RGSi) variant of Gαi2 (RGSi Gαi2) display a spontaneous
Gary R Ten Eyck et al.
Pharmacology, biochemistry, and behavior, 199, 173068-173068 (2020-11-05)
This ethopharmacological investigation comprised a long-term field study that examined the function of serotonergic and vasotonergic systems in territoriality. Adult territorial and non-territorial (silent) male coquí frogs (Eleutherodactylus coqui) were injected (IP) with either arginine vasotocin (AVT) or one of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service