Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

150134

Sigma-Aldrich

Isoquinoline-1-carboxylic acid

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$140.00
25 G
$364.00

$140.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$140.00
25 G
$364.00

About This Item

Empirical Formula (Hill Notation):
C10H7NO2
CAS Number:
Molecular Weight:
173.17
Beilstein:
129177
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$140.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

99%

mp

164 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1nccc2ccccc12

InChI

1S/C10H7NO2/c12-10(13)9-8-4-2-1-3-7(8)5-6-11-9/h1-6H,(H,12,13)

InChI key

XAAKCCMYRKZRAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isoquinoline-1-carboxylic acid was used in the preparation of 4-cyano-3-ethoxy-1-hydroxy-5,6,7,8-tetrahydroisoquinoline[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Knoevenagel condensation of 2-carbethoxycyclohexanone and malononitrile: Synthesis of 4-cyano-3-ethoxy-1-hydroxy-5, 6, 7, 8-tetrahydroisoquinoline, an isomer of the abnormal product.
Kasturi TR and Sharma VK.
Tetrahedron, 31(6), 527-531 (1975)
Jennifer A Jacobsen et al.
Journal of medicinal chemistry, 54(2), 591-602 (2010-12-30)
Fragment-based lead design (FBLD) has been used to identify new metal-binding groups for metalloenzyme inhibitors. When screened at 1 mM, a chelator fragment library (CFL-1.1) of 96 compounds produced hit rates ranging from 29% to 43% for five matrix metalloproteases

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service