Key Documents
SYNA851922
Synthware U-shaped drying tube
joint: ST/NS 19/22
Synonym(s):
Synthware A851922, Drying tube adapter
Select a Size
1 EA
$55.20
$55.20
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$55.20
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$55.20
Please contact Customer Service for Availability
material
colorless tube
glass tube
Joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A851922
General description
With joint. Cotton is usually used to contain drying agent.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)