Documents
SYNB262914
Synthware bubbler with PTFE stopcock
top joint: ST/NS 29/42, bottom joint: ST/NS 14/20
Synonym(s):
Synthware B262914, mineral oil bubbler
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
bottom joint: ST/NS14/20
joint: ST/NS top29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware B262914
Related Categories
General description
With small ball joints to prevent mineral oil being sucked back under sudden negative pressure in the system.
Legal Information
Synthware is a trademark of Kemtech America Inc.