Documents
SLW4513/13M
Pyrex® Culture tube cap inserts
thread size 13 mm
Synonym(s):
Pyrex, SciLabware 4513/13M
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
thread size
13 mm
manufacturer/tradename
4513/13M
SciLabware
General description
The in vitro cultivation of animal and plant cells in a nutrient medium is an essential part of today′s research in the field of microbiology. These cultivation approaches can take place at different scales - the vessels are selected accordingly. For test runs, rather small vessels such as culture tubes are used.
Features and Benefits
• Manufactured from borosilicate glass 3.3• High resistance to attack from water reduces leaching of contaminants which can cause pH changes• PBT screwcaps with inert PTFE lined rubber disc• Cap can withstand repeated dry heat and steam sterilisation cycles.
Other Notes
Insert Only
Legal Information
PYREX is a registered trademark of Corning, Inc.