SLW3152/04AWC
Pyrex® Volumetric flask, heavy duty, class A, amber, works certified, USP/ISO/DIN tolerances
capacity 5 mL
Synonym(s):
Pyrex, SciLabware 3152/04AWC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass flask
Joint
joint ST/NS neck10/19
feature
accuracy class A (ISO 1042 and DIN 12664, United States Pharmacopoeia (USP),conformity ′H′ certified)
packaging
pkg of 5 ea
manufacturer/tradename
SciLabware 3152/04AWC
Overall H
70 mm
capacity
5 mL
Looking for similar products? Visit Product Comparison Guide
General description
Class A, amber, works certified
- Complies with ISO 1042 and DIN 12664
- Calibrated to the tolerances required by the United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards
- Each flask engraved with individual serial number and supplied with certificate of calibration traceable to National standards
- Manufactured from borosilicate glass with an amber coating on exterior surfaces to facilitate use with light sensitive solutions
- Conformity ‘H′ certified
- Polyethylene stopper
- White enamel inscriptions
Features and Benefits
• Complies with ISO 1042 and DIN 12664• Manufactured from borosilicate glass 3.3• Calibrated to the tolerances required by the United States Pharmacopoeia (USP) which are tighter than those of ISO/DIN standards• With an amber coating on exterior surfaces to facilitate use with light sensitive solutions• Inscriptions in white enamel• Each flask engraved with individual serial number and supplied with certificate of calibration traceable to National standards• Batch certificate available to download online.• With Polyethylene stopper
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service