Skip to Content
MilliporeSigma
All Photos(1)

Documents

QBD10931

Sigma-Aldrich

m-dPEG®36-MAL

>95% (HPLC)

Synonym(s):

mPEG36-MAL, mPEG-maleimide, mPEG1500-maleimide, mPEG36-maleimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C80H154N2O39
Molecular Weight:
1768.07
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

m-dPEG36-MAL is a methyl-terminated, thiol-reactive, monodispersed PEGylation product designed for surface and biomolecule modification. This single molecular weight PEG reagent has a discrete chain length of 116 atoms spanning a distance of 132.4 Å. One end of the molecule terminates in a maleimido-propionic acid group, which is conjugated to the dPEG spacer by an amide bond. The maleimide moiety reacts with sulfhydryls to form a thioether bond. dPEG36 is a monodisperse equivalent of the polymeric PEG1500. Modification of cysteine residues on peptides, proteins, and other sulfhydryl-containing molecules with m-dPEG36-MAL will increase hydrophilicity and decrease immunogenicity.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service