Skip to Content
MilliporeSigma
All Photos(1)

Documents

JKA3124

Sigma-Aldrich

mPEG2K

average Mn 2,000, Maleimide, thiol reactive

Synonym(s):

mPEG-MAL, Methoxy-PEG-Maleimide, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)nCH2CH2NHCOCH2CH2C4H2O2N
UNSPSC Code:
12162002
NACRES:
NA.23

product name

mPEG2K-MAL, average Mn 2,000

form

solid

mol wt

average Mn 2,000

reaction suitability

reagent type: chemical modification reagent
reactivity: thiol reactive

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Methoxy PEG Maleimide. Selective for thiol groups on cystein side chains; reacts at pH 5.0-6.7

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu, W., et al.
Antiviral Research, 108 (2014)
Pradyumna Kedarisetti et al.
Biomedical optics express, 11(11), 6211-6230 (2020-12-08)
The presence of circulating tumor cells (CTCs) in a patient's bloodstream is a hallmark of metastatic cancer. The detection and analysis of CTCs is a promising diagnostic and prognostic strategy as they may carry useful genetic information from their derived

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service