Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Q205

Sigma-Aldrich

p-Quaterphenyl

98%

Synonym(s):

Benzerythrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4C6H4C6H5
CAS Number:
Molecular Weight:
306.40
Beilstein:
1912745
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

98%

bp

428 °C/18 mmHg (lit.)

mp

>300 °C (lit.)

λmax

298 nm

SMILES string

c1ccc(cc1)-c2ccc(cc2)-c3ccc(cc3)-c4ccccc4

InChI

1S/C24H18/c1-3-7-19(8-4-1)21-11-15-23(16-12-21)24-17-13-22(14-18-24)20-9-5-2-6-10-20/h1-18H

InChI key

GPRIERYVMZVKTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingliang Zhang et al.
Journal of chromatography. A, 1365, 148-155 (2014-09-25)
One-pot synthesis of surface-confined ionic liquid functionalized silica spheres was proposed using N-(3-aminopropyl)imidazole, γ-isopropyltriethoxysilane and 1-bromooctadecane as starting materials. The surface modification of the silica spheres was successful with a high surface density of octadecylimidazolium, enabling the utilization of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service