MilliporeSigma
  • A novel urea-functionalized surface-confined octadecylimidazolium ionic liquid silica stationary phase for reversed-phase liquid chromatography.
A novel urea-functionalized surface-confined octadecylimidazolium ionic liquid silica stationary phase for reversed-phase liquid chromatography.
Journal of chromatography. A (2014-09-25)
Mingliang Zhang, Ting Tan, Zhan Li, Tongnian Gu, Jia Chen, Hongdeng Qiu
ABSTRACT
One-pot synthesis of surface-confined ionic liquid functionalized silica spheres was proposed using N-(3-aminopropyl)imidazole, γ-isopropyltriethoxysilane and 1-bromooctadecane as starting materials. The surface modification of the silica spheres was successful with a high surface density of octadecylimidazolium, enabling the utilization of this new urea-functionalized ionic liquid-grafted silica material as stationary phase for high-performance liquid chromatography in reversed-phase mode. The long aliphatic chain combined with the multiple polar group embedded in the ligands imparted the new stationary phase fine selectivity towards PAH isomers and polar aromatics and higher affinity for phenolic compounds. The unique features of the new material, especially the effect of the urea group on the retention were elucidated by mathematic modeling.
Sigma-Aldrich
p-Terphenyl, ≥99.5% (HPLC)
Supelco
Chrysene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Biphenyl, PESTANAL®, analytical standard
Sigma-Aldrich
Biphenyl, ≥99%
Supelco
Chrysene, analytical standard
Supelco
p-Terphenyl, analytical standard
Sigma-Aldrich
p-Terphenyl, suitable for scintillation, ≥98.5% (HPLC)
Chrysene, BCR®, certified reference material
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Biphenyl, ReagentPlus®, 99.5%
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
1-(3-Aminopropyl)imidazole, ≥97%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Supelco
Urea, analytical standard
Urea, European Pharmacopoeia (EP) Reference Standard