Documents
MT51192107
Mettler Toledo LogStraight fingerprint reader
About This Item
UNSPSC Code:
41115600
NACRES:
NB.79
manufacturer/tradename
Mettler-Toledo 51192107
General description
No more user names to enter, no more cumbersome password management. With LogStraight, the biometric fingerprint reader, the operators simply put their finger onto the reader and the DM density meter or RM refractometer knows immediately who they are.