Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JKA13019

Sigma-Aldrich

mPEG12-Azide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)11CH2CH2N3
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

liquid

mol wt

average Mn 12,000

reaction suitability

reagent type: chemical modification reagent
reaction type: click chemistry

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

General description

Monodisperse PEG. Azido group can be reduced to the amine. Azido can reactive with alkyne through click chemistry.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Protocols

Analysis of gene expression data requires a stable reference or loading control. This reference is usually one or more reference genes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service