Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JKA12018

Sigma-Aldrich

Propionic Acid-PEG12-propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCOCH2CH2O(CH2CH2O)12CH2CH2COOH
UNSPSC Code:
12352106
NACRES:
NA.23

form

solid

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service